Tisk…

Methyl vanillate 98%

BLDPBD6734-10GEA 285 CZK
BLDPBD6734-10G BLDPBD6734-100G BLDPBD6734-25G BLDPBD6734-500G BLDPBD6734-5G
Methyl vanillate 98%
Methyl vanillate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₁₀O₄
MW: 182,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008438
CAS číslo: 3943-74-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR