Tisk…

4-(2-Carboxyethyl)phenylboronic acid 98%

BLDPBD6757-1GEA 1800 CZK
BLDPBD6757-1G BLDPBD6757-250MG BLDPBD6757-5G
4-(2-Carboxyethyl)phenylboronic acid 98%
4-(2-Carboxyethyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₉H₁₁BO₄
MW: 193,99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318119
CAS číslo: 166316-48-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR