Tisk…

2,6-Dibromo-4-tert-butylphenol 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD68056-25GEA 3030 CZK
BLDPBD68056-25G BLDPBD68056-5G BLDPBD68056-10G BLDPBD68056-250MG BLDPBD68056-0.025G BLDPBD68056-1G
2,6-Dibromo-4-tert-butylphenol 95%
2,6-Dibromo-4-tert-butylphenol

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₁₂Br₂O
MW: 308,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00152389
CAS číslo: 98-22-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR