Tisk…

2,6-Difluoro-4-iodoaniline 97%

BLDPBD68272-25GEA 2000 CZK
BLDPBD68272-0.025G BLDPBD68272-25G BLDPBD68272-100G BLDPBD68272-250MG BLDPBD68272-1G BLDPBD68272-10G BLDPBD68272-5G
2,6-Difluoro-4-iodoaniline 97%
2,6-Difluoro-4-iodoaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₆H₄F₂IN
MW: 255,0039 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07774189
CAS číslo: 141743-49-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR