Fmoc-N-amido-PEG10-Acid 97%
|
|
BLDPBD683436-1GEA
10860
CZK
BLDPBD683436-1G
BLDPBD683436-100MG
BLDPBD683436-5G
BLDPBD683436-250MG
BLDPBD683436-10G
Fmoc-N-amido-PEG10-Acid 97%
Fmoc-N-amido-PEG10-Acid
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₃₈H₅₇NO₁₄ MW: 751,857 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28122954 CAS číslo: 2101563-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...