Tisk…

2,6-Dimethylisonicotinic acid 98%

BLDPBD68486-250MGEA 684 CZK
BLDPBD68486-250MG BLDPBD68486-1G BLDPBD68486-100MG BLDPBD68486-5G
2,6-Dimethylisonicotinic acid 98%
2,6-Dimethylisonicotinic acid

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₉NO₂
MW: 151,163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03731176
CAS číslo: 54221-93-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR