Tisk…

8-Boc-2,8-Diazaspiro[4.5]decane 98%

BLDPBD68680-10GEA 32610 CZK
BLDPBD68680-10G BLDPBD68680-25G BLDPBD68680-100MG BLDPBD68680-250MG BLDPBD68680-1G BLDPBD68680-5G
8-Boc-2,8-Diazaspiro[4.5]decane 98%
8-Boc-2,8-Diazaspiro[4.5]decane

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₁₃H₂₄N₂O₂
MW: 240,342 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09608078
CAS číslo: 236406-39-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR