Tisk…

4-(2-Hydroxyethyl)-2-methoxyphenol 95%

BLDPBD6923-50MGEA 3990 CZK
BLDPBD6923-50MG BLDPBD6923-10MG
4-(2-Hydroxyethyl)-2-methoxyphenol 95%
4-(2-Hydroxyethyl)-2-methoxyphenol

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₁₂O₃
MW: 168,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002903
CAS číslo: 2380-78-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR