2-(p-Toluenesulphonyl)ethanol 98%
|
|
BLDPBD6924-250MGEA
314
CZK
BLDPBD6924-250MG
BLDPBD6924-10G
BLDPBD6924-100MG
BLDPBD6924-5G
BLDPBD6924-25G
BLDPBD6924-1G
2-(p-Toluenesulphonyl)ethanol 98%
2-(p-Toluenesulphonyl)ethanol
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₉H₁₂O₃S MW: 200,2600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00041757 CAS číslo: 22381-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...