Tisk…

2-Acetamidothiazole 96%

BLDPBD69387-25GEA 2400 CZK
BLDPBD69387-25G BLDPBD69387-5G
2-Acetamidothiazole 96%
2-Acetamidothiazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₅H₆N₂OS
MW: 142,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016892
CAS číslo: 2719-23-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR