Tisk…

N,N-Dimethyl 3-borono-4-methylbenzenesulphonamide 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD6987-1GEA 6190 CZK
BLDPBD6987-1G BLDPBD6987-250MG BLDPBD6987-5G
N,N-Dimethyl 3-borono-4-methylbenzenesulphonamide 98%
N,N-Dimethyl 3-borono-4-methylbenzenesulphonamide

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₉H₁₄BNO₄S
MW: 243,0900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08056363
CAS číslo: 871332-99-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR