Tisk…

(2S,3aS,7aS)-2-Carboxyoctahydroindole 98%

BLDPBD7040-10GEA 1170 CZK
BLDPBD7040-10G BLDPBD7040-1G BLDPBD7040-100G BLDPBD7040-5G BLDPBD7040-25G
(2S,3aS,7aS)-2-Carboxyoctahydroindole 98%
(2S,3aS,7aS)-2-Carboxyoctahydroindole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₉H₁₅NO₂
MW: 169,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782125
CAS číslo: 80875-98-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR