Methyl 2-amino-5-chloronicotinate 95%
|
|
BLDPBD70417-1GEA
998
CZK
BLDPBD70417-1G
BLDPBD70417-250MG
BLDPBD70417-5G
BLDPBD70417-100MG
Methyl 2-amino-5-chloronicotinate 95%
Methyl 2-amino-5-chloronicotinate
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Vzorec:
C₇H₇ClN₂O₂ MW: 186,596 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09031678 CAS číslo: 50735-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...