2-Bromo-4,5-dimethoxybenzonitrile 98%
|
|
BLDPBD71706-100MGEA
998
CZK
BLDPBD71706-100MG
BLDPBD71706-5G
BLDPBD71706-1G
BLDPBD71706-250MG
2-Bromo-4,5-dimethoxybenzonitrile 98%
2-Bromo-4,5-dimethoxybenzonitrile
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₉H₈BrNO₂ MW: 242,0693 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02256761 CAS číslo: 109305-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...