Tisk…

2-Bromo-3-hydroxy-6-methylpyridine 97%

BLDPBD72072-5GEA 3000 CZK
BLDPBD72072-5G BLDPBD72072-250MG BLDPBD72072-25G BLDPBD72072-1G BLDPBD72072-10G
2-Bromo-3-hydroxy-6-methylpyridine 97%
2-Bromo-3-hydroxy-6-methylpyridine

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₆H₆BrNO
MW: 188,022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06655956
CAS číslo: 23003-35-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR