Tisk…

2-Bromoethyl phenyl sulphide 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD72180-5GEA 1370 CZK
BLDPBD72180-5G BLDPBD72180-1G BLDPBD72180-25G BLDPBD72180-100G
2-Bromoethyl phenyl sulphide 98%
2-Bromoethyl phenyl sulphide

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₈H₉BrS
MW: 217,12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00040826
CAS číslo: 4837-01-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR