Tisk…

2-Chloro-N,N-diisobutylacetamide 95%

BLDPBD73801-25GEA 9980 CZK
BLDPBD73801-25G BLDPBD73801-5G BLDPBD73801-1G
2-Chloro-N,N-diisobutylacetamide 95%
2-Chloro-N,N-diisobutylacetamide

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₀H₂₀ClNO
MW: 205,725 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040035
CAS číslo: 5326-82-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR