Tisk…

4-Methyl-3-(trifluoromethyl)benzoic acid 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD7412-5GEA 428 CZK
BLDPBD7412-5G BLDPBD7412-10G BLDPBD7412-1G
4-Methyl-3-(trifluoromethyl)benzoic acid 98%
4-Methyl-3-(trifluoromethyl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₇F₃O₂
MW: 204,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631592
CAS číslo: 261952-01-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR