Tisk…

1-Cyclopentenylboronic acid pinacol ester 98%

BLDPBD74236-100GEA 14970 CZK
BLDPBD74236-100G BLDPBD74236-1G BLDPBD74236-25G BLDPBD74236-5G BLDPBD74236-10G
1-Cyclopentenylboronic acid pinacol ester 98%
1-Cyclopentenylboronic acid pinacol ester

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₁₁H₁₉BO₂
MW: 194,0784 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669391
CAS číslo: 287944-10-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR