Tisk…

2-Ethynylthiophene 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD74755-25GEA 8640 CZK
BLDPBD74755-25G BLDPBD74755-5G BLDPBD74755-10G BLDPBD74755-0.025G BLDPBD74755-1G
2-Ethynylthiophene 95%
2-Ethynylthiophene

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Vzorec: C₆H₄S
MW: 108,1610 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03425129
CAS číslo: 4298-52-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR