Tisk…

5-Chloro-7-methoxythieno[3,2-b]pyridine 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD750622-250MGEA 2260 CZK
BLDPBD750622-250MG BLDPBD750622-100MG BLDPBD750622-1G BLDPBD750622-5G
5-Chloro-7-methoxythieno[3,2-b]pyridine 97%
5-Chloro-7-methoxythieno[3,2-b]pyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₈H₆ClNOS
MW: 199,657 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10000846
CAS číslo: 90690-91-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR