Tisk…

rac-AntPhos 97%

BLDPBD756263-10GEA 32120 CZK
BLDPBD756263-10G BLDPBD756263-5G BLDPBD756263-100MG BLDPBD756263-1G BLDPBD756263-250MG BLDPBD756263-25G
rac-AntPhos 97%
rac-AntPhos

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₂₅H₂₃OP
MW: 370,423 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30748199
CAS číslo: 1268693-24-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR