Tisk…

Propargyl-PEG3-NHS ester 97%

BLDPBD760153-5GEA 17160 CZK
BLDPBD760153-5G BLDPBD760153-250MG BLDPBD760153-1G BLDPBD760153-100MG
Propargyl-PEG3-NHS ester 97%
Propargyl-PEG3-NHS ester

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₄H₁₉NO₇
MW: 313,303 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23726610
CAS číslo: 1428629-71-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR