Tisk…

Hydroxy-PEG2-CH2CO2t-Bu 95%

BLDPBD760155-5GEA 9520 CZK
BLDPBD760155-5G BLDPBD760155-1G BLDPBD760155-250MG BLDPBD760155-100MG
Hydroxy-PEG2-CH2CO2t-Bu 95%
Hydroxy-PEG2-CH2CO2t-Bu

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₀H₂₀O₅
MW: 220,263 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25959245
CAS číslo: 149299-82-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR