Tisk…

2-Mercaptopyridine-N-oxide 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD76277-100GEA 5930 CZK
BLDPBD76277-100G BLDPBD76277-25G BLDPBD76277-5G
2-Mercaptopyridine-N-oxide 98%
2-Mercaptopyridine-N-oxide

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₅H₅NOS
MW: 127,17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006196
CAS číslo: 1121-31-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR