Tisk…

Methyl-3-formylbenzoate 98%

BLDPBD7630-25GEA 5080 CZK
BLDPBD7630-25G BLDPBD7630-100G BLDPBD7630-1G BLDPBD7630-10G BLDPBD7630-5G BLDPBD7630-250MG
Methyl-3-formylbenzoate 98%
Methyl-3-formylbenzoate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₉H₈O₃
MW: 164,1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00189379
CAS číslo: 52178-50-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR