Zanamivir monohydrate 98%
|
|
BLDPBD763215-250MGEA
2680
CZK
BLDPBD763215-250MG
BLDPBD763215-100MG
Zanamivir monohydrate 98%
Zanamivir monohydrate
Storage condition: Inert atmosphere, 2-8°C
Vzorec:
C₁₂H₂₀N₄O₇·H₂O MW: 350,33 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03791324 CAS číslo: 1260601-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...