Tisk…

2-Bromobenzaldehyde ethylene acetal 98% (by GC)

Dodavatel: BLD PHARMATECH GMBH

BLDPBD7661-100GEA 3650 CZK
BLDPBD7661-100G BLDPBD7661-5G BLDPBD7661-1G BLDPBD7661-10G BLDPBD7661-25G
2-Bromobenzaldehyde ethylene acetal 98% (by GC)
2-Bromobenzaldehyde ethylene acetal

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₉H₉BrO₂
MW: 229,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00155124
CAS číslo: 34824-58-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR