Tisk…

2-Chloro-5-formylphenylboronic acid 97%

BLDPBD8034-10GEA 3340 CZK
BLDPBD8034-10G BLDPBD8034-250MG BLDPBD8034-5G BLDPBD8034-25G BLDPBD8034-1G BLDPBD8034-100G
2-Chloro-5-formylphenylboronic acid 97%
2-Chloro-5-formylphenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₆BClO₃
MW: 184,3847 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06656269
CAS číslo: 1150114-78-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR