Tisk…

3-Benzyloxy-2,6-difluorophenylboronic acid 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD80467-25GEA 10210 CZK
BLDPBD80467-25G BLDPBD80467-5G BLDPBD80467-250MG BLDPBD80467-1G
3-Benzyloxy-2,6-difluorophenylboronic acid 98%
3-Benzyloxy-2,6-difluorophenylboronic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₃H₁₁BF₂O₃
MW: 264,0325 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06200705
CAS číslo: 870718-07-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR