Tisk…

2-Fluoro-m-toluic acid 97%

BLDPBD8065-250MGEA 257 CZK
BLDPBD8065-250MG BLDPBD8065-10G BLDPBD8065-100G BLDPBD8065-5G BLDPBD8065-1G BLDPBD8065-25G
2-Fluoro-m-toluic acid 97%
2-Fluoro-m-toluic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₇FO₂
MW: 154,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042504
CAS číslo: 315-31-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR