3-(Hydroxymethyl)phenylboronic acid 98%
|
|
BLDPBD8123-5GEA
285
CZK
BLDPBD8123-5G
BLDPBD8123-100G
BLDPBD8123-25G
BLDPBD8123-500G
BLDPBD8123-10G
3-(Hydroxymethyl)phenylboronic acid 98%
3-(Hydroxymethyl)phenylboronic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Vzorec:
C₇H₉BO₃ MW: 151,96 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01317846 CAS číslo: 87199-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...