3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt 98%
|
|
BLDPBD82487-500GEA
13770
CZK
BLDPBD82487-500G
BLDPBD82487-5G
BLDPBD82487-100G
BLDPBD82487-25G
BLDPBD82487-0.025G
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt 98%
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₆H₃Cl₂NaO₄S MW: 265,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00009798 CAS číslo: 54970-72-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...