Tisk…

3,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD82882-25GEA 7100 CZK
BLDPBD82882-25G BLDPBD82882-5G BLDPBD82882-100G BLDPBD82882-250MG BLDPBD82882-10G BLDPBD82882-1G
3,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 97%
3,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₁₁H₁₉BN₂O₂
MW: 222,0918 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663852
CAS číslo: 857530-80-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR