Tisk…

3-Amino-4-bromo-1-methyl-1H-pyrazole 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD84126-25GEA 11490 CZK
BLDPBD84126-25G BLDPBD84126-10G BLDPBD84126-1G BLDPBD84126-100G BLDPBD84126-250MG BLDPBD84126-5G
3-Amino-4-bromo-1-methyl-1H-pyrazole 97%
3-Amino-4-bromo-1-methyl-1H-pyrazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₄H₆BrN₃
MW: 176,02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00663065
CAS číslo: 146941-72-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR