Tisk…

3-Aminobenzoic acid hydrochloride 95+%

BLDPBD84414-25GEA 9720 CZK
BLDPBD84414-25G BLDPBD84414-1G BLDPBD84414-5G
3-Aminobenzoic acid hydrochloride 95+%
3-Aminobenzoic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₈ClNO₂
MW: 173,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012973
CAS číslo: 15151-51-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR