Tisk…

3-Bromo-1H-1,2,4-triazole 98%

BLDPBD84863-5GEA 627 CZK
BLDPBD84863-5G BLDPBD84863-10G BLDPBD84863-500G BLDPBD84863-25G BLDPBD84863-100G BLDPBD84863-1G
3-Bromo-1H-1,2,4-triazole 98%
3-Bromo-1H-1,2,4-triazole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₂H₂BrN₃
MW: 147,96 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09055371
CAS číslo: 7343-33-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR