Tisk…

Methyl-3-bromo-2-methylbenzoate 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD84969-5GEA 428 CZK
BLDPBD84969-5G BLDPBD84969-25G BLDPBD84969-10G BLDPBD84969-100G BLDPBD84969-1G BLDPBD84969-500G BLDPBD84969-1000G
Methyl-3-bromo-2-methylbenzoate 98%
Methyl-3-bromo-2-methylbenzoate

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₉H₉BrO₂
MW: 229,0706 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09031772
CAS číslo: 99548-54-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR