Tisk…

3-Bromo-2-thiophenecarbonitrile 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD85426-250MGEA 285 CZK
BLDPBD85426-250MG BLDPBD85426-5G BLDPBD85426-1G
3-Bromo-2-thiophenecarbonitrile 97%
3-Bromo-2-thiophenecarbonitrile

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₅H₂BrNS
MW: 188,05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00068173
CAS číslo: 18791-98-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR