N-Fmoc-D-valine 99%
|
|
BLDPBD8588-25GEA
656
CZK
BLDPBD8588-25G
BLDPBD8588-500G
BLDPBD8588-5G
BLDPBD8588-0.025G
BLDPBD8588-1G
BLDPBD8588-100G
BLDPBD8588-10G
N-Fmoc-D-valine 99%
N-Fmoc-D-valine
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Vzorec:
C₂₀H₂₁NO₄ MW: 339,39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00062953 CAS číslo: 84624-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...