Tisk…

6-Hydroxynicotinonitrile 98%

BLDPBD86310-250MGEA 428 CZK
BLDPBD86310-250MG BLDPBD86310-10G BLDPBD86310-5G BLDPBD86310-25G BLDPBD86310-100MG BLDPBD86310-1G
6-Hydroxynicotinonitrile 98%
6-Hydroxynicotinonitrile

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₆H₄N₂O
MW: 120,109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06656489
CAS číslo: 95891-30-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR