3-Methoxy-N,N-dimethylaniline 95%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD86487-5GEA
599
CZK
BLDPBD86487-5G
BLDPBD86487-100G
BLDPBD86487-25G
3-Methoxy-N,N-dimethylaniline 95%
3-Methoxy-N,N-dimethylaniline
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Vzorec:
C₉H₁₃NO MW: 151,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00051779 CAS číslo: 15799-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...