3-Fluoro-4-methoxybenzylamine 97%
|
|
BLDPBD86823-1GEA
912
CZK
BLDPBD86823-1G
BLDPBD86823-10G
BLDPBD86823-0.025G
BLDPBD86823-25G
BLDPBD86823-250MG
BLDPBD86823-5G
3-Fluoro-4-methoxybenzylamine 97%
3-Fluoro-4-methoxybenzylamine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Vzorec:
C₈H₁₀FNO MW: 155,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04116360 CAS číslo: 123652-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...