Tisk…

3-Fluoroazetidine hydrochloride 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD86930-10GEA 3450 CZK
BLDPBD86930-10G BLDPBD86930-1G BLDPBD86930-5G BLDPBD86930-250MG
3-Fluoroazetidine hydrochloride 97%
3-Fluoroazetidine hydrochloride
Vzorec: C3H7ClFN
MW: 111,55 g/mol
MDL Number: MFCD05663715
CAS číslo: 617718-46-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR