Tisk…

3-Mercaptobenzoic acid 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD87643-10GEA 6700 CZK
BLDPBD87643-10G BLDPBD87643-25G BLDPBD87643-5G BLDPBD87643-250MG BLDPBD87643-1G
3-Mercaptobenzoic acid 97%
3-Mercaptobenzoic acid

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₆O₂S
MW: 154,1863 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00238636
CAS číslo: 4869-59-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR