Tisk…

3-Methoxy-4-nitrobenzaldehyde 95%

BLDPBD87725-100MGEA 1090 CZK
BLDPBD87725-100MG BLDPBD87725-1G BLDPBD87725-5G BLDPBD87725-250MG
3-Methoxy-4-nitrobenzaldehyde 95%
3-Methoxy-4-nitrobenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₈H₇NO₄
MW: 181,1455 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08703367
CAS číslo: 80410-57-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR