Tisk…

3-Methyl-1H-1,2,4-triazole 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD87891-1GEA 456 CZK
BLDPBD87891-1G BLDPBD87891-10G BLDPBD87891-25G BLDPBD87891-100G BLDPBD87891-250MG BLDPBD87891-5G
3-Methyl-1H-1,2,4-triazole 98%
3-Methyl-1H-1,2,4-triazole

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₃H₅N₃
MW: 83,0900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00233931
CAS číslo: 7170-01-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR