Tisk…

3-Methylhippuric acid 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD88243-1GEA 285 CZK
BLDPBD88243-1G BLDPBD88243-5G
3-Methylhippuric acid 98%
3-Methylhippuric acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₁₁NO₃
MW: 193,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044399
CAS číslo: 27115-49-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR