Tisk…

N-Cbz-L-phenylalaninol 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD8885-100GEA 3200 CZK
BLDPBD8885-100G BLDPBD8885-10G BLDPBD8885-1G BLDPBD8885-25G BLDPBD8885-5G
N-Cbz-L-phenylalaninol 98%
N-Cbz-L-phenylalaninol

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₇H₁₉NO₃
MW: 285,3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00191138
CAS číslo: 6372-14-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR