Tisk…

3-Picoline-N-oxide 98%

BLDPBD88975-5GEA 285 CZK
BLDPBD88975-5G BLDPBD88975-25G BLDPBD88975-500G BLDPBD88975-100G
3-Picoline-N-oxide 98%
3-Picoline-N-oxide

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₆H₇NO
MW: 109,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006203
CAS číslo: 1003-73-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR